Skip to Content
Merck
All Photos(1)

Key Documents

45491

Supelco

Fentin acetate

PESTANAL®, analytical standard

Synonym(s):

Triphenyltin acetate, Acetoxytriphenylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SnOCOCH3
CAS Number:
Molecular Weight:
409.07
Beilstein:
4146107
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)O[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.C2H4O2.Sn/c3*1-2-4-6-5-3-1;1-2(3)4;/h3*1-5H;1H3,(H,3,4);/q;;;;+1/p-1

InChI key

WDQNIWFZKXZFAY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Lin et al.
American journal of nephrology, 13(2), 124-128 (1993-01-01)
Three patients who developed acute nephropathy following ingestion of triphenyltin acetate (TPTA) are described. All of them had significant proteinuria, azotemia, and polyuria. Mild neurological manifestations in all patients were also noted. Hematuria and pyuria were noted in 1 severely
C Nebbia et al.
Journal of toxicology and environmental health. Part A, 56(6), 433-447 (1999-03-30)
The in vitro effects of the organotin (OT) compound triphenyltin acetate (TPTA) on cytochrome P-450 content and functions were investigated in liver microsomes from untreated, phenobarbital (PB)- or beta-naphthoflavone- (betaNAF) pretreated rats. At a concentration of 0.5 mM, TPTA caused
I Roessink et al.
Ecotoxicology (London, England), 15(3), 267-293 (2006-03-25)
Floodplain lakes in the Rhine-Meuse delta of the Netherlands vary considerably in levels of sediment-bound toxicants. Microcosm experiments were done to compare the ecological impact of the fungicide triphenyltin acetate (TPT) between test systems with clean or polluted sediments (10
M Dacasto et al.
Schweizer Archiv fur Tierheilkunde, 136(3), 111-115 (1994-01-01)
Twenty-eight immature male rabbits were divided into four experimental groups and administered a standard diet containing respectively 0, 15, 75 and 150 ppm of triphenyltin acetate (TPTA) for 70 days. The highest dose group showed a decrease in erythrocyte count
U S Attahiru et al.
Veterinary and human toxicology, 33(5), 499-502 (1991-10-01)
The effects of a moderate but prolonged exposure of rats to tri-n-butyltin acetate (TBTA) and triphenyltin acetate (TPTA), as might be encountered as residues in yam peel diets, were investigated in rats. These subchronic toxicity studies (16, 8, or 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service