45489
Fenson
PESTANAL®, analytical standard
Synonym(s):
4-Chlorophenyl benzenesulfonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
Clc1ccc(OS(=O)(=O)c2ccccc2)cc1
InChI
1S/C12H9ClO3S/c13-10-6-8-11(9-7-10)16-17(14,15)12-4-2-1-3-5-12/h1-9H
InChI key
SPJOZZSIXXJYBT-UHFFFAOYSA-N
Related Categories
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Recommended products
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
>212.0 °F
Flash Point(C)
> 100 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bulletin of environmental contamination and toxicology, 104(5), 568-574 (2020-04-24)
Water quality degradation by organochlorine pesticides and potentially toxic elements is of worldwide concern. This research explores groundwater conditions, regarding organochlorine pesticides and potentially toxic elements, in Hopelchen, Campeche, which is located in the buffer zone of the Calakmul Biosphere
Gas chromatographic determination of organochlorine pesticides; contamination of Dicofol, Fenson, and Tetradifon in fish and natural waters of a wet area beside the Mediterranean Sea.
Bulletin of environmental contamination and toxicology, 36(2), 211-218 (1986-02-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service