Skip to Content
Merck
All Photos(2)

Key Documents

45448

Supelco

Dimethametryn

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$103.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$103.00

About This Item

Empirical Formula (Hill Notation):
C11H21N5S
CAS Number:
Molecular Weight:
255.38
Beilstein:
528188
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$103.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)C(C)C)nc(SC)n1

InChI

1S/C11H21N5S/c1-6-12-9-14-10(13-8(4)7(2)3)16-11(15-9)17-5/h7-8H,6H2,1-5H3,(H2,12,13,14,15,16)

InChI key

IKYICRRUVNIHPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Matsui et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 51(3-4), 329-337 (2005-04-27)
In the prediction of time-series concentrations of herbicides in river water with diffuse-pollution hydrological models, farming schedules (the dates of herbicide application and drainage of irrigation water from rice paddies) greatly affect the runoff behavior of the herbicides. For large
Acute toxicity of pesticides to Gobius sp., Palaemonetes africanus, and Desmocaris trispimosa.
A G Ebere et al.
Bulletin of environmental contamination and toxicology, 49(4), 588-592 (1992-10-01)
Kunihiko Fujii et al.
Pest management science, 63(3), 254-260 (2007-01-25)
A novel bacterial strain FJ1117YT was isolated from an enrichment culture with the herbicide simetryn. The isolate was capable of degrading the herbicide supplied as the sole sulfur source in an aquatic batch culture. The strain FJ1117YT was identified as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service