Skip to Content
Merck
All Photos(1)

Key Documents

45431

Supelco

Dichlobenil

PESTANAL®, analytical standard

Synonym(s):

2,6-Dichlorobenzonitrile, Dichlobenil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CN
CAS Number:
Molecular Weight:
172.01
Beilstein:
1909167
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

143-146 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cccc(Cl)c1C#N

InChI

1S/C7H3Cl2N/c8-6-2-1-3-7(9)5(6)4-10/h1-3H

InChI key

YOYAIZYFCNQIRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlobenil is classified under the nitrile group of herbicides.[1]

Application

Dichlobenil may be used as an analytical standard for the determination of dichlobenil in:
  • Cereals[2] and dry fruits[3] by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and gas chromatography coupled to tandem mass spectrometry (GC-MS/MS) with multiple reaction monitoring (MRM) detection.
  • Food samples by GC-MS/MS with MRM detection.[4]
  • Grape seed extracts by GC coupled to triple quadrupole mass spectrometry (QqQ-MS/MS) equipped with electron ionization (EI) and MRM detection.[5]
  • Wheat flour samples by magnetic solid-phase extraction (MSPE) along with high performance liquid chromatography-diode array detection (DAD).[6]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid Multi-Residue Analysis of Herbicides with Endocrine-Disrupting Properties in Environmental Water Samples Using Ultrasound-Assisted Dispersive Liquid-Liquid Microextraction and Gas Chromatography-Mass Spectrometry.
Shi XZ, et al.
Chromatographia, 1-13 (2018)
Simultaneous determination of bifenox, dichlobenil and diclofop methyl by hollow carbon nanospheres enhanced magnetic carboxylic multi-walled carbon nanotubes.
Li WK, et al.
Analytica Chimica Acta, 1011, 40-49 (2018)
Multiresidue analysis of over 200 pesticides in cereals using a QuEChERS and gas chromatography?tandem mass spectrometry-based method.
He Z, et al.
Food Chemistry, 169, 372-380 (2015)
Hugo Mélida et al.
Phytochemistry, 71(14-15), 1684-1689 (2010-07-20)
Cultured maize cells habituated to grow in the presence of the cellulose synthesis inhibitor dichlobenil (DCB) have a modified cell wall in which the amounts of cellulose are reduced and the amounts of arabinoxylan increased. This paper examines the contribution
Tomoyuki Tsuchiyama et al.
Journal of chromatography. A, 1524, 233-245 (2017-10-19)
In the multi-residue analysis of pesticides using GC-MS, the quantitative results are adversely affected by a phenomenon known as the matrix effect. Although the use of matrix-matched standards is considered to be one of the most practical solutions to this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service