Skip to Content
Merck
All Photos(2)

Key Documents

45383

Supelco

Dalapon-methyl

PESTANAL®, analytical standard

Synonym(s):

Methyl 2,2-dichloropropionate, Dalapon-methyl

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$84.40

$84.40


0 left in stock

Request a Bulk Order

Select a Size

Change View
250 MG
$84.40

About This Item

Linear Formula:
CH3CCl2COOCH3
CAS Number:
Molecular Weight:
157.00
Beilstein:
1751091
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$84.40


0 left in stock

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(C)(Cl)Cl

InChI

1S/C4H6Cl2O2/c1-4(5,6)3(7)8-2/h1-2H3

InChI key

RDYUJBOUMXIDPY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service