Skip to Content
Merck
All Photos(1)

Key Documents

448516

Sigma-Aldrich

2-Pentanone

ReagentPlus®, ≥99%, purified by redistillation

Synonym(s):

Methyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
506058
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
grade:
reagent
bp:
101-105 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)

grade

reagent

vapor pressure

27 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

941 °F

purified by

redistillation

expl. lim.

8.2 %

dilution

(for general lab use)

refractive index

n20/D 1.39 (lit.)

bp

101-105 °C (lit.)

mp

−78 °C (lit.)

solubility

water: soluble 72.6 g/L at 20 °C

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)=O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward rational design of stable, supported metal catalysts for aqueous-phase processing: Insights from the hydrogenation of levulinic acid.
Abdelrahman OA, et al.
J. Catal., 329, 10-21 (2015)
On the dissociation of the 2-pentanone ion studied by threshold photoelectron photoion coincidence spectroscopy.
Kercher JP, et al.
International Journal of Mass Spectrometry, 249, 403-411 (2006)
Infinite-dilution binary diffusion coefficients of 2-propanone, 2-butanone, 2-pentanone, and 3-pentanone in CO2 by the Taylor dispersion technique from 308.15 to 328.15 K in the pressure range from 8 to 35 MPa.
Funazukuri T, et al.
International Journal of Thermophysics, 21(6), 1279-1290 (2000)
Atmospheric chemistry of 2-pentanone and 2-heptanone.
Atkinson R, et al.
Environmental Science & Technology, 34(4), 623-631 (2000)
Phase liquid-vapor equilibria and thermodynamic properties of solutions of n-propanol-aliphatic ketones.
Suntsov YK, et al.
Russian Journal of Physical Chemistry A, 89(6), 926-930 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service