Skip to Content
Merck
All Photos(2)

Key Documents

44722

Supelco

trans,trans-Farnesyl pyrophosphate ammonium salt

analytical standard

Synonym(s):

(E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt, Farnesyl diphosphate ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$922.00

$922.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$922.00

About This Item

Empirical Formula (Hill Notation):
C15H28O7P2 · xNH3
Molecular Weight:
382.33 (free acid basis)
Beilstein:
2482197
UNSPSC Code:
85151701
NACRES:
NA.24

$922.00


Available to ship on07 April 2025Details


Request a Bulk Order

Quality Level

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

N.C\C(C)=C\CC\C(C)=C\CC\C(C)=C\COP(O)(=O)OP(O)(O)=O

InChI

1S/C15H28O7P2.H3N/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-21-24(19,20)22-23(16,17)18;/h7,9,11H,5-6,8,10,12H2,1-4H3,(H,19,20)(H2,16,17,18);1H3/b14-9+,15-11+;

InChI key

CPYJTMLHKIWGDF-NDHHSALASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

NMR-Internal Standard: not specified dimethylsulfone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service