Skip to Content
Merck
All Photos(1)

Key Documents

44292

Supelco

(+)-Garcinia acid

analytical standard

Synonym(s):

(−)-Hydroxycitric acid lactone, (2S,3S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid, Garcinia lactone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$356.00

$356.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$356.00

About This Item

Empirical Formula (Hill Notation):
C6H6O7
CAS Number:
Molecular Weight:
190.11
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$356.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

OC(=O)[C@H]1OC(=O)C[C@@]1(O)C(O)=O

InChI

1S/C6H6O7/c7-2-1-6(12,5(10)11)3(13-2)4(8)9/h3,12H,1H2,(H,8,9)(H,10,11)/t3-,6+/m1/s1

InChI key

PFHZIWAVXDSFTB-CVYQJGLWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gopalakrishnan Saroja Seethapathy et al.
Scientific reports, 8(1), 10561-10561 (2018-07-14)
Garcinia L. (Clusiaceae) fruits are a rich source of (-)-hydroxycitric acid, and this has gained considerable attention as an anti-obesity agent and a popular weight loss food supplement. In this study, we assessed adulteration of morphologically similar samples of Garcinia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service