Skip to Content
Merck
All Photos(2)

Key Documents

44291

Supelco

Dimethyl isophthalate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1912251
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6.8 (vs air)

vapor pressure

93.7 mmHg ( 208 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

64-68 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COC(=O)c1cccc(c1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-4-3-5-8(6-7)10(12)14-2/h3-6H,1-2H3

InChI key

VNGOYPQMJFJDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl isophthalate belongs to the class of phthalates, which are widely used as plasticizers to make hard plastics more flexible. They are utilized in a variety of products including cosmetics, detergents, lubricating oils, solvents, polychlorinated biphenyl substitutes, carriers in pesticide formulations, building products such as flooring, sheeting, films, personal care products, medical devices, food packaging and toys. Exposure to phthalates is inevitable due to its prevalence in most of the consumer products. Thus, there is continued investigation and ongoing research to determine the potential health hazards of these group of chemicals.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Dimethyl isophthalate may be used as an internal standard for determining, dimethyl-tere-phthalate (DMtP), dimethyl-phthalate (DMP), diethyl phthalate (DEP) in soil samples and biosolids using gas-chromatography coupled to mass spectrometry (GC-MS). It may also be used as an analytical reference standard for the quantification of the analyte in complex matrices including plastics, cosmetics, and environmental samples using gas chromatography coupled to positive chemical ionization mass spectrometry (GC-CIMS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

343.4 °F - closed cup

Flash Point(C)

173 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phthalate esters by positive chemical ionization MS with retention-time locked GC
Jankowski JS, et al.
LCGC North America, 20(2), 142-151 (2002)
Analysis of 4-nonylphenols, phthalates, and polychlorinated biphenyls in soils and biosolids
Gibson R, et al.
Chemosphere, 61(9), 1336-1344 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service