Skip to Content
Merck
All Photos(1)

Key Documents

43908

Sigma-Aldrich

Divinylbenzene

technical, mixture of isomers, ~80% (GC)

Synonym(s):

Diethenylbenzene, Vinylstyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH=CH2)2
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12162002

vapor density

4.5 (vs air)

vapor pressure

0.9 mmHg ( 30 °C)

grade

technical

Assay

~80% (GC)

autoignition temp.

878 °F

contains

~0.1% 4-tert-butylcatechol as stabilizer

expl. lim.

0.7 %, 75 °F
6.5 %, 130 °F

refractive index

n20/D 1.561 (lit.)
n20/D 1.573

bp

195 °C (lit.)

density

0.919 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccc(C=C)cc1

InChI

1S/C10H10/c1-3-9-7-5-6-8-10(9)4-2/h3-8H,1-2H2

InChI key

MYRTYDVEIRVNKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

residual mainly 3- and 4-ethylvinylbenzene

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trishna Saigal et al.
Langmuir : the ACS journal of surfaces and colloids, 29(12), 3999-4007 (2013-03-02)
Multiarm star copolymers with approximately 460 poly(ethylene oxide) (PEO) arms that have a degree of polymerization N = 45 were synthesized via atom transfer radical polymerization (ATRP) of PEO-methacrylate macromonomers in the presence of divinyl benzene cross-linkers. These are an
Toine Hillenaar et al.
Investigative ophthalmology & visual science, 52(5), 2136-2146 (2010-11-13)
To report intra- and interinstrument calibration methods for corneal backscatter analysis by in vivo confocal microscopy. Applicability of two reference standards was evaluated for corneal backscatter calibration. Repeated measurements of four concentrations of AMCO Clear (GFS Chemicals, Inc., Powell, OH)
Yi-Jie Cheng et al.
Journal of chromatography. A, 1218(42), 7640-7647 (2011-07-05)
In this work, a series of poly(divinylbenzene-alkyl methacrylate) monolithic stationary phases, which were prepared by single step in situ polymerization of divinylbenzene and various alkyl methacrylates (butyl-, octyl-, lauryl- or stearyl methacrylate), were developed as separation columns of nine common
Rainer Koeck et al.
The Analyst, 138(17), 5089-5098 (2013-06-27)
Monolithic capillary columns were prepared by thermally initiated free radical copolymerization of N-vinylcarbazole (NVC) and 1,4-divinylbenzene (DVB) within the confines of 200 and 100 μm i.d. fused silica capillaries. The reaction was carried out under the influence of inert micro-(toluene)
Fatemeh S Mirnaghi et al.
Journal of chromatography. A, 1261, 91-98 (2012-07-25)
This work presents the development and evaluation of biocompatible polyacrylonitrile-polystyrene-divinylbenzene (PAN-PS-DVB) and polyacrylonitrile-phenylboronic acid (PAN-PBA) coatings for automated 96-blades (thin-film) solid phase microextraction (SPME) system, using high performance liquid chromatography (HPLC) coupled with tandem mass spectrometry (MS/MS). The SPME condition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service