Skip to Content
Merck
All Photos(1)

Key Documents

43503

Supelco

Potassium bromide

≥99.9995% (metals basis)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Assay

≥99.9995% (metals basis)

impurities

≤10 mg/kg total nitrogen (N)

mp

734 °C (lit.)

anion traces

bromate (BrO3-): ≤10 mg/kg
chloride (Cl-): ≤500 mg/kg
iodide (I-): ≤200 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤0.02 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.5 mg/kg
Ca: ≤0.1 mg/kg
Cd: ≤0.005 mg/kg
Ce: ≤0.005 mg/kg
Co: ≤0.005 mg/kg
Cr: ≤0.005 mg/kg
Cu: ≤0.005 mg/kg
Fe: ≤0.01 mg/kg
Hg: ≤0.005 mg/kg
La: ≤0.005 mg/kg
Li: ≤0.5 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.005 mg/kg
Na: ≤5 mg/kg
Ni: ≤0.005 mg/kg
Pb: ≤0.005 mg/kg
Sc: ≤0.005 mg/kg
Sm: ≤0.005 mg/kg
Sr: ≤0.05 mg/kg
Tl: ≤0.01 mg/kg
Y: ≤0.005 mg/kg
Zn: ≤0.01 mg/kg

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for removal of peripheral membrane proteins.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dariusz Wawro et al.
International journal of molecular sciences, 12(11), 7286-7300 (2011-12-17)
This paper describes a method for preparing chitosan fibers modified with hydroxyapatite (HAp), tricalcium phosphate (β-TCP), and HAp/β-TCP nanoparticles. Fiber-grade chitosan derived from the northern shrimp (Pandalus borealis) and nanoparticles of tricalcium phosphate (β-TCP) and hydroxyapatite (HAp) suspended in a
Mamta J Patel et al.
Journal of cellular biochemistry, 106(2), 306-316 (2009-01-07)
Bone loss due to osteoporosis or disuse such as in paraplegia or microgravity is a significant health problem. As a treatment for osteoporosis, brief exposure of intact animals or humans to low magnitude and high frequency (LMHF) mechanical loading has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service