Skip to Content
Merck
All Photos(1)

Key Documents

42891

Supelco

Hyperforin (dicyclohexylammonium) salt

analytical standard

Synonym(s):

Hyp-DCHA, Hyperforin-DCHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52O4·C12H23N
CAS Number:
Molecular Weight:
718.10
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water (Karl Fischer)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)C(=O)[C@@]34C(O)=C(C\C=C(\C)C)C(=O)[C@@](C\C=C(\C)C)(C[C@H](C\C=C(/C)C)[C@@]3(C)CC\C=C(\C)C)C4=O

InChI

1S/C35H52O4.C12H23N/c1-22(2)13-12-19-33(11)27(16-14-23(3)4)21-34(20-18-25(7)8)30(37)28(17-15-24(5)6)31(38)35(33,32(34)39)29(36)26(9)10;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h13-15,18,26-27,38H,12,16-17,19-21H2,1-11H3;11-13H,1-10H2/t27-,33+,34+,35-;/m0./s1

InChI key

ZUYCAROZOLUHJY-DNSWOBEMSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort.
Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found it to have neuroprotective effects against Alzheimer′s disease (AD) neuropathology, including the ability to disassemble amyloid-β aggregates in vitro and improve spatial memory in vivo.
Stable salt of hyperforin.
Stable salt of hyperforin. Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. Activation of TRPC6 induces the entry of sodium and calcium into the cell which causes inhibition of monoamine reuptake. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found other neurological effects, effects on inflammation, as well as antibacterial, antitumoral and antiangiogenic effects.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service