Skip to Content
Merck
All Photos(2)

Key Documents

42643

Supelco

(−)-Huperzine A

analytical standard

Synonym(s):

(−)-Selagine, Kimpukan A

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$337.00

$337.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$337.00

About This Item

Empirical Formula (Hill Notation):
C15H18N2O
CAS Number:
Molecular Weight:
242.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$337.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Assay

≥98.0% (HPLC)

optical activity

[α]/D -153±5°, c = 0.5 in methanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC3=C(C=CC(=O)N3)[C@@](N)(CC(C)=C1)C2=CC

InChI

1S/C15H18N2O/c1-3-11-10-6-9(2)8-15(11,16)12-4-5-14(18)17-13(12)7-10/h3-6,10H,7-8,16H2,1-2H3,(H,17,18)/b11-3+/t10-,15+/m0/s1

InChI key

ZRJBHWIHUMBLCN-YQEJDHNASA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Huperzine A, a sesquiterpene alkaloid isolated from the Chinese herb Huperzia serrata. Its mode of action involves the strong inhibition of acetylcholinesterase (AChE).[1][2]

Application

(−)-Huperzine A may be used as a reference standard in the determination of the analyte in plasma samples using high-performance liquid chromatography (HPLC) with fluorescence detection.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Huperzine A in rat plasma by high-performance liquid chromatography with a fluorescence detector.
Yue P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 44(1), 309-312 (2007)
Andrea Zangara
Pharmacology, biochemistry, and behavior, 75(3), 675-686 (2003-08-05)
Huperzine A (HupA), extracted from a club moss (Huperzia serrata), is a sesquiterpene alkaloid and a powerful and reversible inhibitor of acetylcholinesterase (AChE). It has been used in China for centuries for the treatment of swelling, fever and blood disorders.
Rui Wang et al.
Acta pharmacologica Sinica, 27(1), 1-26 (2005-12-21)
Huperzine A (HupA), a novel alkaloid isolated from the Chinese herb Huperzia serrata, is a potent, highly specific and reversible inhibitor of acetylcholinesterase(AChE). Compared with tacrine, donepezil, and rivastigmine, HupA has better penetration through the blood-brain barrier, higher oral bioavailability
Yongrui Du et al.
Scientific reports, 8(1), 6166-6166 (2018-04-20)
Acetylcholine (ACh) in the ovary and its actions were linked to survival of human granulosa cells in vitro and improved fertility of rats in vivo. These effects were observed upon experimental blockage of the ACh-degrading enzyme (ACH esterase; ACHE), by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service