Skip to Content
Merck
All Photos(1)

Key Documents

42376

Millipore

Violet Red Bile Lactose Agar, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Crystal violet-Neutral red-Bile Agar, Vegiton, VRB-Agar, Vegitone, Vegitone Violet Red Bile Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Product 42376 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.0 g/L
crystal violet, 0.002 g/L
lactose, 10.0 g/L
neutral red, 0.03 g/L
peptone (vegetable), 7.0 g/L
sodium chloride, 5.0 g/L
synthetic detergent, 1.5 g/L
yeast extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Salmonella spp.
selective and differential for coliforms

Application

Violet Red Bile Agar Vegitone is a selective media used for the detection and enumeration of coliform organisms from water and food. This medium contains no animal derived peptone and bile salt.

Preparation Note

Suspend 38.53 g in 1000 ml distilled water. Heat with stirring to boiling to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 45°C and pour into sterile petri plates containing the inoculum.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service