Skip to Content
Merck
All Photos(1)

Key Documents

42363

Supelco

(±)-Enterolactone

analytical standard

Synonym(s):

Enterolactone, trans-α,β-Bis(3-hydroxybenzyl)butyrolactone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$670.00

$670.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$670.00

About This Item

Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
Beilstein:
4699604
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$670.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

suitability

conforms to structure for Proton NMR spectrum

format

neat

storage temp.

2-8°C

SMILES string

Oc1cccc(C[C@@H]2COC(=O)[C@H]2Cc3cccc(O)c3)c1

InChI

1S/C18H18O4/c19-15-5-1-3-12(8-15)7-14-11-22-18(21)17(14)10-13-4-2-6-16(20)9-13/h1-6,8-9,14,17,19-20H,7,10-11H2/t14-,17+/m1/s1

InChI key

HVDGDHBAMCBBLR-PBHICJAKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Enterolactone is the major human metabolite of dietary sesamin (lignan from sesame seeds). A phytoestrogen, it shows an inverse correlation with risk of breast cancer in one study.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bárbara Socas-Rodríguez et al.
Analytical and bioanalytical chemistry, 409(18), 4467-4477 (2017-06-07)
Within this study, a new method enabling monitoring of various estrogenic substances potentially occurring in milk and dairy products was proposed. Groups of compounds fairly differing in physico-chemical properties and biological activity were analyzed: four natural estrogens, four synthetic estrogens
Bárbara Socas-Rodríguez et al.
Journal of chromatography. A, 1496, 58-67 (2017-04-02)
In this work, a simple and fast methodology has been validated and applied for the analysis of a group of 22 estrogenic compounds including eight phytoestrogens (i.e. daidzein, enterodiol, glycitein, enterolactone, genistein, formononetin, prunetin, biochanin A), six mycotoxins (β-zearalanol, β-zearalenol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service