Skip to Content
Merck
All Photos(1)

Key Documents

42277

Supelco

(±)-2-Octanol

analytical standard

Synonym(s):

Hexyl methyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$98.40

$98.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$98.40

About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719325
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$98.40


Estimated to ship on29 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.426

density

0.819 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Zhang et al.
Journal of agricultural and food chemistry, 62(33), 8302-8307 (2014-07-30)
Nonvolatile compounds play important roles in the quality of alcoholic beverages. In our previous work, a type of cyclooctapeptide lichenysin was newly identified in Chinese strong-aroma type liquor. In this work, it was found that lichenysin could selectively affect aroma
Anzi Wang et al.
Journal of chromatography. A, 1374, 216-223 (2014-12-06)
Fabrication technologies for microelectromechanical systems (MEMS) allow miniaturization of conventional benchtop gas chromatography (GC) to portable, palm-sized microfabricated GC (μGC) devices, which are suitable for on-site chemical analysis and remote sensing. The separation performance of μGC systems, however, has not
Pankaj K Sethi et al.
Journal of pharmacological and toxicological methods, 70(1), 106-111 (2014-06-15)
A simple, reliable procedure was developed to measure binding of pyrethroid insecticides to total proteins and lipoproteins of rat and human plasma. The extent of binding of (14)C-labeled deltamethrin (DLM), cis-permethrin (CIS) and trans-permethrin (TRANS) was quantified by a 3-step
Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of
Zuobing Xiao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 978-979, 122-130 (2014-12-30)
A simple, rapid and solvent-free multi-residue method has been developed and applied to confirm and quantify a series of volatile compounds in five cherry wines by gas chromatography coupled with mass spectrometry (GC-MS). Four parameters (e.g., coating material of fiber

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service