Skip to Content
Merck
All Photos(1)

Key Documents

42258

Supelco

(S)-ar-Turmerone

analytical standard

Synonym(s):

(S)-2-Methyl-6-(4-methylphenyl)-2-hepten-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O
CAS Number:
Molecular Weight:
216.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

Pricing and availability is not currently available.

grade

analytical standard

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](CC(=O)\C=C(\C)C)c1ccc(C)cc1

InChI

1S/C15H20O/c1-11(2)9-15(16)10-13(4)14-7-5-12(3)6-8-14/h5-9,13H,10H2,1-4H3/t13-/m0/s1

InChI key

NAAJVHHFAXWBOK-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-ar-Turmerone is the major constituent of turmeric oil derived from turmeric[1], which can find pharmaceutical applications due to its antibacterial, anti-carcinogenic, insect-repellent, anti-oxidant and anti-fungal properties.[2][1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chae Hee Hong et al.
Planta medica, 68(6), 545-547 (2002-07-03)
Beta-turmerone and ar-turmerone, sesquiterpenoids isolated from the rhizome of Curcuma zedoaria, inhibited lipopolysaccharide (LPS)-induced prostaglandin E 2 production in cultured mouse macrophage cell RAW 264.7 in a dose-dependent manner (IC 50 = 7.3 microM for beta-turmerone; IC 50 = 24.0
Tan Yonggang et al.
Human vaccines & immunotherapeutics, 8(10), 1416-1424 (2012-10-26)
The aim of this work is to evaluate the effects of purified aromatic-turmerone (ar-turmerione, AR) on murine dendritic cells (DCs). These impacts of AR on DCs from bone marrow derived DCs(BMDCs) were assessed with use of conventional scanning electron microscopy
N Y Qin et al.
Journal of pharmaceutical and biomedical analysis, 43(2), 486-492 (2006-08-26)
Curcuma longa (Zingiberaceae) is a native plant of southern Asia and is cultivated extensively throughout the warmer parts of the world. Jianghuang and Yujin are rhizome and tuberous root of C. longa, respectively, which were traditionally used as two Chinese
Preparation of turmeric oil-loaded chitosan-alginate biopolymeric nanocapsules
Lertsutthiwong P, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 29, 856-860 (2009)
H S Lee
Bioresource technology, 97(12), 1372-1376 (2005-08-23)
The antiplatelet activities of Curcuma longa L. rhizome-derived materials were measured using a platelet aggregometer and compared with those of aspirin as antiplatelet agent. The active constituent from the rhizome of Curcuma longa L. was isolated and characterized as ar-turmerone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service