Skip to Content
Merck
All Photos(2)

Key Documents

42102

Supelco

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide

for chiral derivatization, LiChropur, ≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$279.00

$279.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$279.00

About This Item

Linear Formula:
FC6H2(NO2)2NH[CHCH(CH3)2]CONH2
CAS Number:
Molecular Weight:
300.24
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

$279.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

derivatization grade ((chiral))
for chiral derivatization

Quality Level

Assay

≥98.0% (sum of enantiomers, HPLC)
≥98.0%

optical activity

[α]20/D +14.0±1.5°, c = 2% in acetone

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

mp

173-177 °C (lit.)
173-177 °C

storage temp.

2-8°C

SMILES string

CC(C)[C@H](Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C11H13FN4O5/c1-5(2)10(11(13)17)14-7-3-6(12)8(15(18)19)4-9(7)16(20)21/h3-5,10,14H,1-2H3,(H2,13,17)/t10-/m0/s1

InChI key

ZTFFRKNPAXXEBL-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide [L-FDVA] is a derivatizing chiral agent.[1]

Application

L-FDVA may be used as derivatizing agent for amino acids in the isolation and identification of microcystins.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral derivatizing agent (chiral Sanger′s Reagent) for determining the enantiomeric purity of amino acids with HPLC[2]; This analogue of Marfey′s reagent can also be used with amines[3][4]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography-electrospray ionisation-mass spectrometry based method for the simultaneous determination of algal and cyanobacterial toxins in phytoplankton from marine waters and lakes followed by tentative structural elucidation of microcystins.
Dahlmann, Jens, Wes R. Budakowski, and Bernd Luckas
Journal of Chromatography A, 994, 45-57 (2003)
H. Bruckner et al.
Journal of Chromatography A, 555, 81-81 (1991)
M. Calmes et al.
Tetrahedron Asymmetry, 4, 2437-2437 (1993)
K. Shimada et al.
Journal of Liquid Chromatography, 16, 3311-3311 (1993)
Eric J Dimise et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(40), 15311-15316 (2008-10-04)
Bacteria belonging to the order Actinomycetales have proven to be an important source of biologically active and often therapeutically useful natural products. The characterization of orphan biosynthetic gene clusters is an emerging and valuable approach to the discovery of novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service