Skip to Content
Merck
All Photos(1)

Key Documents

41135

Supelco

Periodic table mix 2 for ICP

TraceCERT®, 17 elements, 10 mg/L

Synonym(s):

Periodic table mix 2 for ICP, 17 elements

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$623.00

$623.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$623.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$623.00


Available to ship on01 April 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

10 mg/L

technique(s)

AAS: suitable
ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Periodic table mix 2 can be used as a calibration standard in the quantification of multi-element concentrations in a variety of water, beverage, and biofluid samples by inductively coupled plasma mass spectrometry method (ICP-MS).[1]

Analysis Note

10 mg/L: Au, Ge, Hf, Ir, Mo, Nb, Pd, Pt, Re, Rh, Ru, Sb, Sn, Ta, Ti, W, Zr in 5%hydrochloric acid and 1% hydrofluoric acid (contains HNO3 traces)

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Z Martín-González et al.
Nutrients, 12(2) (2020-02-09)
Conjugated linoleic acid (CLA) is a dietary supplement that has been shown to improve obesity. However, some authors have associated high doses of CLA supplementation with liver impairment and insulin resistance. The aim of this study was to assess whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service