Skip to Content
Merck
All Photos(1)

Key Documents

41062

Supelco

Sodium tetrapropylborate

for GC derivatization, LiChropur, ≥85.0% (T)

Synonym(s):

NaBPr4

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$346.00

$346.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 G
$346.00

About This Item

CAS Number:
Beilstein:
4007550
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$346.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥85.0% (T)

form

solid

quality

LiChropur

reaction suitability

reagent type: catalyst
core: boron

reagent type: derivatization reagent
reaction type: Alkylations

technique(s)

gas chromatography (GC): suitable

SMILES string

CCC[B-](CCC)(CCC)CCC.[Na+]

InChI

1S/C12H28B.Na/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;/q-1;+1

InChI key

VKBNWVIDTHWBAZ-UHFFFAOYSA-N

General description

Sodium tetrapropylborate (NaBPr4) belongs to the class of tetraalkylborates, which are commonly used as derivatization agents to accomplish the multielement speciation of organometallic compounds on account of their ability to react with elements such as mercury, lead and tin. NaBPr4 is preferred because it eliminates the need for organic solvents as required in Grignard reactions, also resulting in reduced analysis time.[1][2][3][4][5]

Application

Sodium tetrapropylborate may be used as a derivatization agent to enhance the sensitivity of methylmercury in fish,[1] organolead compounds in water and road dust samples[2] and mercury species in sea water samples[3] prior to their determination using microwave digestion with gas chromatography-mass spectrometry (GC-MS), GC coupled to microwave-induced plasma atomic-emission spectrometry (MIP-AES) and GC with inductively coupled plasma (ICP) MS, respectively.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ propylation using sodium tetrapropylborate as a fast and simplified sample preparation for the speciation analysis of organolead compounds using GC-MIP-AES
Heisterkamp M and Adams FC
Journal of Analytical Atomic Spectrometry, 14(9), 1307-1311 (1999)
Comparison of two derivatization reagents for the simultaneous determination of organolead and organomanganese compounds using solid-phase microextraction followed by gas chromatography with atomic emission detection
Pe?alver R, et al.
Talanta, 87(8), 268-275 (2011)
Determination of methylmercury in fish using focused microwave digestion following by Cu2+ addition, sodium tetrapropylborate derivatization, n-heptane extraction, and gas chromatography-mass spectrometry
Chen SS, et al.
Journal of Chromatography A, 1024(1-2), 209-215 (2004)
Mercury speciation analysis in sea water by solid phase microextraction-gas chromatography-inductively coupled plasma mass spectrometry using ethyl and propyl derivatization. Matrix effects evaluation
Bravo-Sanchez LR, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 59(1), 59-66 (2004)
A comparison of alkyl derivatization methods for speciation of mercury based on solid phase microextraction gas chromatography with furnace atomization plasma emission spectrometry detection.
Grinberg P, et al.
Journal of Analytical Atomic Spectrometry, 18(8), 902-909 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service