Skip to Content
Merck
All Photos(1)

Key Documents

40251

Sigma-Aldrich

N,N-Dimethylformamide ethylene acetal

technical, ≥90% (NT)

Synonym(s):

2-Dimethylamino-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2
CAS Number:
Molecular Weight:
117.15
Beilstein:
773813
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (NT)

impurities

some dimethylformamide

refractive index

n20/D 1.429

bp

142-144 °C (lit.)

density

1.015 g/mL at 20 °C (lit.)

SMILES string

CN(C)C1OCCO1

Other Notes

Review

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.F. Abdulla et al.
Tetrahedron, 35, 1675-1675 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service