Skip to Content
Merck
All Photos(3)

Documents

391557

Sigma-Aldrich

Oil Blue N

Dye content 96 %

Synonym(s):

Solvent Blue 14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30N2O2
CAS Number:
Molecular Weight:
378.51
Colour Index Number:
61555
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 96%

mp

112-114 °C (lit.)

solubility

acetone: 1%, turbid, blue to very deep blue

λmax

596 nm
644 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCCCNc1ccc(NCCCCC)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C24H30N2O2/c1-3-5-9-15-25-19-13-14-20(26-16-10-6-4-2)22-21(19)23(27)17-11-7-8-12-18(17)24(22)28/h7-8,11-14,25-26H,3-6,9-10,15-16H2,1-2H3

InChI key

RHGBRYSELHPAFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In animal tissues, fatty substances are stained dark blue by oil blue N when supersaturated solutions in dilute isopropanol are used.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oil Blue N or Na as a Fat Stain for Animal Histology.
Lillie R D
Stain Technology, 20, 7-9 (1945)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service