Skip to Content
Merck
All Photos(1)

Key Documents

38948

Millipore

Cronobacter screening broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

CSB, Cronobacter Selective Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Product 38948 is not currently sold in your country. Contact Technical Service

Agency

according to ISO 22964:2017

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 4 mg/L
meat extract, 3.0 g/L
peptone, 10.0 g/L
sodium chloride, 5.0 g/L
sucrose, 10.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

Cronobacter spp.

Application

An enrichment broth for the detection of Cronobacter (formerly E. sakazakii) from food and environmental samples.

Preparation Note

Dissolve 28 g in 1000 ml of distilled water. Sterilize by autoclaving at 121°C for 15 minutes. Cool to approximately 50°C, and aseptically add the contents of 1 vial of Vancomycin Supplement (Cat. No. 75423) reconstituted as directed. Mix well and aseptically dispense into sterile containers.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service