Skip to Content
Merck
All Photos(2)

Key Documents

38874

Supelco

Pseudoerythromycin A enol ether

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$886.00

$886.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$886.00

About This Item

Empirical Formula (Hill Notation):
C37H65NO12
CAS Number:
Molecular Weight:
715.91
UNSPSC Code:
41116107
NACRES:
NA.24

$886.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CC[C@@H](O)[C@@](C)(O)C1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)CC(C)=C(O4)[C@@H]1C

InChI

1S/C37H65NO12/c1-14-25(39)37(10,43)32-20(4)28-18(2)16-36(9,50-28)31(49-34-27(40)24(38(11)12)15-19(3)45-34)21(5)29(22(6)33(42)48-32)47-26-17-35(8,44-13)30(41)23(7)46-26/h19-27,29-32,34,39-41,43H,14-17H2,1-13H3/t19-,20+,21+,22-,23+,24+,25-,26+,27-,29+,30+,31-,32-,34+,35-,36-,37-/m1/s1

InChI key

NMIWBQUQCOMGHJ-FYFYGOHNSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Greenwood et al.
Gastroenterology, 106(3), 624-628 (1994-03-01)
Erythromycin (EM-A) and some of its analogues stimulate gastrointestinal smooth muscle contractions. Because gastroesophageal reflux disease (GERD) in humans is in part caused by a reduction in lower esophageal sphincter (LES) pressure, the aim of this study was to investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service