Skip to Content
Merck
All Photos(1)

Documents

38399

Sigma-Aldrich

Diisopropylfluorophosphate

≥97.0% (GC)

Synonym(s):

DFP, DIFP, Diisopropyl phosphorofluoridate, Phosphoric acid diisopropyl ester fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]2POF
CAS Number:
Molecular Weight:
184.15
Beilstein:
1723307
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor pressure

0.58 mmHg ( 20 °C)

Assay

≥97.0% (GC)

refractive index

n20/D 1.385 (lit.)

bp

62 °C/9 mmHg (lit.)

mp

−82 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)OP(F)(=O)OC(C)C

InChI

1S/C6H14FO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3

InChI key

MUCZHBLJLSDCSD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent inhibitor of serine proteases such as trypsin and chymotrypsin, and of acetylcholinesterase; also inhibits cathepsin G, cholinesterase, coagulation factor Xa, leucocyte elastase, pancreatic elastase, tissue kallikrein, plasmin, subtilisin, and thrombin. Inhibition of acetylcholinesterase makes this compound especially toxic. Inhibits apoptosis induced by ricin and bacterial toxins.

Analysis Note

Typically used at a concentration of 0.10 mM. A safer alternative inhibitor for serine protease is phenylmethylsulfonyl fluoride (PMSF).

Other Notes

Potent inhibitor of chymotrypsin and other enzymes (e.g. serine hydrolases); Inhibition of carboxypeptidase but not DAP I

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.K. McDonald et al.
Methods in Enzymology, XV(B), 275-275 (1972)
A molecular probe for the highly selective chromogenic detection of DFP, a mimic of Sarin and Soman nerve agents.
Raúl Gotor et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(43), 11994-11997 (2011-09-17)
Yonggang Li et al.
Toxicology and applied pharmacology, 262(2), 194-204 (2012-05-16)
Current medical countermeasures against organophosphate (OP) nerve agents are effective in reducing mortality, but do not sufficiently protect the CNS from delayed brain damage and persistent neurological symptoms. In this study, we examined the efficacy of neuregulin-1 (NRG-1) in protecting
J.A. Cohen et al.
Methods in Enzymology, XI, 686-686 (1967)
A V Terry et al.
Neurotoxicology and teratology, 34(1), 1-8 (2011-10-26)
The acute toxicity of organophosphates (OPs) has been studied extensively; however, much less attention has been given to the subject of repeated exposures that are not associated with overt signs of toxicity (i.e., subthreshold exposures). The objective of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service