Skip to Content
Merck
All Photos(1)

Key Documents

37308

Sigma-Aldrich

(4S,5S)-(−)-4,5-Dihydro-4-methoxymethyl-2-methyl-5-phenyloxazole

purum, ≥97.0% (sum of enantiomers, GC)

Synonym(s):

(4S,5S)-4-Methoxymethyl-2-methyl-5-phenyl-2-oxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2
CAS Number:
Molecular Weight:
205.25
Beilstein:
1214742
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D −115.0±2°, c = 10.5% in chloroform

SMILES string

COC[C@@H]1N=C(C)O[C@H]1c2ccccc2

Other Notes

Chiral acetimidate useful for the preparation of chiral β-substituted acids, aldehydes or alcohols[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Whittaker et al.
Canadian Journal of Chemistry, 63, 2844-2844 (1985)
A.I. Meyers et al.
The Journal of Organic Chemistry, 44, 2250-2250 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service