Recommended Products
Agency
USP/NF
reag. Ph. Eur.
feature
non-bleeding: no
refill: no
application(s)
general analytical
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
Packaging
1 Packing = 1 booklet with 100 strips each
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemical analyses of aqueous extract of Parkia biglobosa fruit husk collected from Northern Ghana.
Scientific Research and Essays, 8, 589-595 (2013)
Remington: The Science and Practice of Pharmacy, 500-500 (2006)
Science Fair Projects: Chemistry, 12-12 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service