Skip to Content
Merck
All Photos(1)

Key Documents

37077

Supelco

Pyribenzoxim

PESTANAL®, analytical standard

Synonym(s):

Benzophenone O-[2,6-bis(4,6-dimethoxypyrimidin-2-yloxy)benzoyl]oxime, Diphenylmethanone O-[2,6-bis[(4,6-dimethoxy-2-pyrimidinyl)oxy]benzoyl]oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H27N5O8
CAS Number:
Molecular Weight:
609.59
Beilstein:
8369592
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(Oc2cccc(Oc3nc(OC)cc(OC)n3)c2C(=O)O\N=C(/c4ccccc4)c5ccccc5)n1

InChI

1S/C32H27N5O8/c1-39-24-18-25(40-2)34-31(33-24)43-22-16-11-17-23(44-32-35-26(41-3)19-27(36-32)42-4)28(22)30(38)45-37-29(20-12-7-5-8-13-20)21-14-9-6-10-15-21/h5-19H,1-4H3

InChI key

OVXMBIVWNJDDSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service