Skip to Content
Merck
All Photos(1)

Key Documents

36925

Supelco

Diallyl phthalate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$59.30

$59.30


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$59.30

About This Item

Linear Formula:
C6H4-1,2-(CO2CH2CH=CH2)2
CAS Number:
Molecular Weight:
246.26
Beilstein:
1880877
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$59.30


Estimated to ship on22 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

8.3 (vs air)

vapor pressure

2.3 mmHg ( 150 °C)

autoignition temp.

725 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.519 (lit.)

bp

165-167 °C/5 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

C=CCOC(=O)c1ccccc1C(=O)OCC=C

InChI

1S/C14H14O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h3-8H,1-2,9-10H2

InChI key

QUDWYFHPNIMBFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beng H Tan et al.
Advances in colloid and interface science, 113(2-3), 111-120 (2005-06-07)
The effect of cross-linked density on the rheological behavior of model pH-responsive microgel systems consisting of methacrylic acid-ethyl acrylate (MAA-EA) cross-linked with di-allyl phthalate (DAP) was examined. Neutralization of acid groups increases the osmotic pressure exerted by counter-ions trapped in
Jin Zhou et al.
Chemosphere, 79(5), 595-604 (2010-03-02)
The contamination of marine ecosystems by endocrine disrupting compounds (EDCs) is of great concern. Protein expression profile maybe a good method to help us understand the molecular mechanisms of EDCs-toxicity to aquatic organisms. In this study, the abalone (Haliotis diversicolor
A Gomiero et al.
Aquatic toxicology (Amsterdam, Netherlands), 78 Suppl 1, S34-S41 (2006-04-25)
Ciphergen ProteinChip Technology is a proteomic tool, used for the discovery of new and sensitive biomarkers. This approach was used to evaluate the protein profile of crabs exposed to various pollutants. Two different exposure experiments were performed: spider crabs (Hyas
D A Eigenberg et al.
Toxicology and applied pharmacology, 86(1), 12-21 (1986-10-01)
In this study we confirmed that diallyl phthalate (DAP) is more hepatotoxic to rats than to mice, and we demonstrated the same species difference in toxicity for allyl alcohol (AA). The data suggest that the toxicity of DAP probably results
Shi-Ai Xu et al.
Ultrasonics sonochemistry, 15(4), 364-369 (2007-10-24)
The bulk polymerization of diallyl phthalate (DAP) was carried out at high temperature (190 degrees C) without using any initiator, and the reaction was stopped before the gelation point in order to get the prepolymer of DAP. The mixture for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service