Skip to Content
Merck
All Photos(1)

Documents

36761

Supelco

3-Chloro-4-methylaniline

PESTANAL®, analytical standard

Synonym(s):

3-Chloro-p-toluidine, 4-Amino-2-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
636511
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.584 (lit.)

bp

237-238 °C (lit.)

mp

24-25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1ccc(N)cc1Cl

InChI

1S/C7H8ClN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3

InChI key

RQKFYFNZSHWXAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randal S Stahl et al.
Journal of agricultural and food chemistry, 50(4), 732-738 (2002-02-07)
A method using a deuterated surrogate of the avicide 3-chloro-p-toluidine hydrochloride (CPTH) was developed to quantify the CPTH residues in the gastrointestinal (GI) tract and breast muscle tissues in birds collected in CPTH-baited sunflower and rice fields. This method increased
Angshuman Bagchi et al.
Computational biology and chemistry, 30(3), 227-232 (2006-05-25)
Microbial redox reactions involving inorganic sulfur compounds in the environment are one of the major reactions of the global sulfur cycle. These reactions are mediated by phylogenetically diverse prokaryotes containing the sulfur oxidizing gene cluster (sox). The sox gene cluster
David A Goldade et al.
Journal of agricultural and food chemistry, 52(26), 8074-8080 (2004-12-23)
Ring-labeled [14C]-3-chloro-4-methylaniline hydrochloride (250 microg per bird) was delivered to 21 red-winged blackbirds (Agelaius phoeniceus) and 21 dark-eyed juncos (Junco hyemalis) via oral gavage, and the distribution and excretion of radioactivity were determined at 15 and 30 min and 1
J C Hurley et al.
Journal of agricultural and food chemistry, 47(7), 2904-2907 (1999-12-20)
Stabilization of the avicide 3-chloro-p-toluidine (CPTH) on rice baits by pseudo latex polymeric coating and beta-cyclodextin inclusion was investigated. When CPTH-treated rice baits were exposed to sunlight, the CPTH formed colored compounds, which exacerbated problems with bait acceptance and efficacy.
[Method of determining p-nitrotoluene, 2-chloro-4-nitrotoluene and 3-chloro-4-methylaniline in the air].
I I Pilenkova et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 43-44 (1980-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service