Skip to Content
Merck
All Photos(1)

Key Documents

36522

Supelco

Quinmerac

PESTANAL®, analytical standard

Synonym(s):

7-Chloro-3-methyl-8-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$191.00

$191.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C11H8ClNO2
CAS Number:
Molecular Weight:
221.64
Beilstein:
8392904
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$191.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cnc2c(C(O)=O)c(Cl)ccc2c1

InChI

1S/C11H8ClNO2/c1-6-4-7-2-3-8(12)9(11(14)15)10(7)13-5-6/h2-5H,1H3,(H,14,15)

InChI key

ALZOLUNSQWINIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Quinmerac is a highly selective herbicide, which belongs to the class of substituted quinolinecarboxylic acids.[1] It can be used in weed control management, particularly for dicotyledonous weeds like cleavers, sugar beet[2], oilseed rape and small grain cereals.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New plant growth regulators: High risk investment?
Rademacher.W and Bucci.T
HortTechnology, 12, 64-67 (2002)
Abscisic acid is a causative factor in the mode of action of the auxinic herbicide quinmerac in cleaver (Galium aparine L.)
Sheltrup F and Grossman K
Journal of Plant Physiology, 147, 118-126 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service