Skip to Content
Merck
All Photos(1)

Key Documents

36465

Supelco

Avenanthramide C

analytical standard

Synonym(s):

2-{[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]amino}-5-hydroxybenzoic acid, N-[3′,4′-Dihydroxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid, Avenanthramide Bc

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$684.00

$684.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$684.00

About This Item

Empirical Formula (Hill Notation):
C16H13NO6
CAS Number:
Molecular Weight:
315.28
Beilstein:
3624543
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$684.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC=C(NC(/C=C/C2=CC(O)=C(O)C=C2)=O)C(C(O)=O)=C1

InChI

1S/C16H13NO6/c18-10-3-4-12(11(8-10)16(22)23)17-15(21)6-2-9-1-5-13(19)14(20)7-9/h1-8,18-20H,(H,17,21)(H,22,23)/b6-2+

InChI key

IDUUXROOZBOOPH-QHHAFSJGSA-N

General description

Avenanthramides are a class of substituted N-cinnamoylanthranilic acids present mainly in oats.[1]

Application

Avenanthramide C may be used as a reference standard for the determination of avenanthramide C in:
  • Oat seeds and seedlings by reversed phase ultra-high performance liquid chromatography equipped with photodiode array deterctor (RP-UHPLC-PDA) and electrospray ionization ion trap mass spectrometry (ESI-IT-MS).[2]
  • Oat-containing products by HPLC combined with triple quadrupole mass spectrometry (TQMS).[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Guadalupe Quiroz Vazquez et al.
Biophysical chemistry, 263, 106391-106391 (2020-05-16)
The misfolding of protein and its assembly into amyloid fibrils with a characteristic β-sheet-rich secondary structure, cause a lot of illnesses. Polyphenols have been extensively studied as a class of amyloid inhibitors, whose effect depends on the position and number
Mass spectrometric characterisation of avenanthramides and enhancing their production by germination of oat (Avena sativa).
de Bruijn WJC, et al.
Food Chemistry, 63(9-10), 419-423 (2018)
Rapid quantitation of avenanthramides in oat-containing products by high-performance liquid chromatography coupled with triple quadrupole mass spectrometry (HPLC-TQMS).
Xie Z, et al.
Food Chemistry, 224(9-10), 280-288 (2017)
Selective and sensitive LC-MS determination of avenanthramides in oats.
Jastrebova J, et al.
Chromatographia, 63(9-10), 419-423 (2006)
Oswaldo Hernandez-Hernandez et al.
Food chemistry, 343, 128408-128408 (2020-11-08)
From a mutagenized oat population, produced by ethyl methanesulfonate mutagenesis, hulled grains from 17 lines with elevated avenanthramide (AVN) content were selected and their AVN structures, concentrations and antioxidant potentials were determined by HPLC-MS2 and HPLC equipped with an on-line

Questions

  1. Hi, can you send us how to dissolve to powder of Avenanthramide C?

    1 answer
    1. This material is not tested for solubility. However, various sources report that this compound is soluble in organic solvents, such as DMSO, alcohols, chloroform, dichloromethane, and ethyl acetate. This information has not been validated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service