Skip to Content
Merck
All Photos(2)

Key Documents

35999

Supelco

Pyrimidifen

PESTANAL®, analytical standard

Synonym(s):

5-Chloro-N-{2-[4-(2-ethoxyethyl)-2,3-dimethylphenoxy]ethyl}-6-ethylpyrimidin-4-amine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$345.00

$345.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$345.00

About This Item

Empirical Formula (Hill Notation):
C20H28ClN3O2
CAS Number:
Molecular Weight:
377.91
Beilstein:
11644658
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$345.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOCCc1ccc(OCCNc2ncnc(CC)c2Cl)c(C)c1C

InChI

1S/C20H28ClN3O2/c1-5-17-19(21)20(24-13-23-17)22-10-12-26-18-8-7-16(9-11-25-6-2)14(3)15(18)4/h7-8,13H,5-6,9-12H2,1-4H3,(H,22,23,24)

InChI key

ITKAIUGKVKDENI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyrimidifen may be used as a reference standard for the determination of the analyte in agricultural products[1] and vegetables or food commodities[2] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation and uncertainty study of a comprehensive list of 160 pesticide residues in multi-class vegetables by liquid chromatography-tandem mass spectrometry.
Kmellar B, et al.
Journal of Chromatography A, 1215(1-2), 37-50 (2008)
Validation of multiresidue screening methods for the determination of 186 pesticides in 11 agricultural products using gas chromatography (GC).
Hirahara Y, et al.
Journal of Health Science, 51(5), 617-627 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service