Skip to Content
Merck
All Photos(1)

Key Documents

35977

Supelco

Tentoxin

analytical standard

Synonym(s):

Cyclo[N-methyl-L-alanyl-L-leucyl-(αZ)-α,β-didehydro-N-methylphenylalanylglycyl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N4O4
CAS Number:
Molecular Weight:
414.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Product 35977 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(C)C[C@@H]1NC(=O)[C@H](C)N(C)C(=O)CNC(=O)C(=C\c2ccccc2)\N(C)C1=O

InChI

1S/C22H30N4O4/c1-14(2)11-17-22(30)26(5)18(12-16-9-7-6-8-10-16)21(29)23-13-19(27)25(4)15(3)20(28)24-17/h6-10,12,14-15,17H,11,13H2,1-5H3,(H,23,29)(H,24,28)/b18-12-/t15-,17-/m0/s1

InChI key

SIIRBDOFKDACOK-LFXZBHHUSA-N

General description

Tentoxin is a phytotoxic tetrapeptide, produced in some Alternaria species.[1] It is known to inhibit cyclic photophosphorylation.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tentoxin may be used as an analytical reference standard for the determination of the analyte in tomato[3] and food matrices[4] by chromatography techniques.

Analysis Note

purity : ≥97.0% (HPLC)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a LC-ESI-MS/MS method for the determination of Alternaria toxins alternariol, alternariol methyl-ether and tentoxin in tomato and tomato-based products.
Rodriguez-Carrasco Y, et al.
Toxins, 8(11), 328-328 (2016)
Inhibition of chloroplast development by tentoxin.
Bennett J.
Phytochemistry, 15(2), 263-265 (1976)
Application of an LC--MS/MS based multi-mycotoxin method for the semi-quantitative determination of mycotoxins occurring in different types of food infected by moulds.
Sulyok M, et al.
Food Chemistry, 119(1), 408-416 (2010)
The sequence and optical configuration of the amino acids in tentoxin.
Koncewicz M, et al.
Biochemical and Biophysical Research Communications, 53(2), 653-658 (1973)
Georg Groth et al.
The Journal of biological chemistry, 277(23), 20117-20119 (2002-04-12)
In contrast to the homologous bacterial and mitochondrial enzymes the chloroplast F(1)-ATPase (CF(1)) is strongly affected by the phytopathogenic inhibitor tentoxin. Based on structural information obtained from crystals of a CF(1)-tentoxin co-complex (Groth, G. (2002) Proc. Natl. Acad. Sci. U.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service