Skip to Content
Merck
All Photos(1)

Key Documents

35867

Sigma-Aldrich

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate

≥98.0% (TLC)

Synonym(s):

3,5-Dichloro-2-hydroxybenzenesulfonic acid, DCHBS, DHBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NaO4S
CAS Number:
Molecular Weight:
265.05
Beilstein:
3640643
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

≥98.0% (TLC)

solubility

H2O: 50 mg/mL, clear, colorless

SMILES string

[Na+].Oc1c(Cl)cc(Cl)cc1S([O-])(=O)=O

InChI

1S/C6H4Cl2O4S.Na/c7-3-1-4(8)6(9)5(2-3)13(10,11)12;/h1-2,9H,(H,10,11,12);/q;+1/p-1

InChI key

NMWCVZCSJHJYFW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions. Component of Trinder reagent for use with peroxidase to measure generation of hydrogen peroxide in automated systems.

Other Notes

Chromogenic substrate for the spectrophotometric assay of hydrogen peroxide[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Wang et al.
Fresenius Journal of Analytical Chemistry, 344, 556-556 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service