Skip to Content
Merck
All Photos(2)

Key Documents

35346

Supelco

Pyraflufen-ethyl

PESTANAL®, analytical standard

Synonym(s):

Ethyl [2-chloro-5-(4-chloro-5-difluoromethoxy-1-methylpyrazol-3-yl)-4-fluorophenoxy]acetate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$289.00

$289.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$289.00

About This Item

Empirical Formula (Hill Notation):
C15H13Cl2F3N2O4
CAS Number:
Molecular Weight:
413.18
Beilstein:
11519423
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$289.00


Estimated to ship on12 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)COc1cc(c(F)cc1Cl)-c2nn(C)c(OC(F)F)c2Cl

InChI

1S/C15H13Cl2F3N2O4/c1-3-24-11(23)6-25-10-4-7(9(18)5-8(10)16)13-12(17)14(22(2)21-13)26-15(19)20/h4-5,15H,3,6H2,1-2H3

InChI key

APTZNLHMIGJTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyraflufen-ethyl is a herbicide which belongs to the phenylpyrazole class.[1] It is used in controlling annual and perennial broadleaf weeds.[1] The mode of action involves inhibiting the enzyme activity of protoporphyrinogen oxidase (Protox).[2]

Application

Pyraflufen-ethyl may be used as a reference standard for the determination of pyraflufen-ethyl herbicide content in fruits and soil samples by high-performance liquid chromatography (HPLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herbicidal activity and characteristics of pyraflufen-ethyl for controlling broad-leaved weeds in cereals.
Tsutomu M, et al.
Journal of pest science, 27(1), 39-46 (2002)
Analysis of pyraflufen-ethyl residues in apples and soil by high-performance liquid chromatography.
Wang H, et al.
International Journal of Environmental Analytical Chemistry, 87(2), 99-109 (2007)
Parthenium hysterophorus L. control in response to pyraflufen-ethyl application.
Fernandez J V, et al.
Crop Protection, 57, 35-37 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service