Skip to Content
Merck
All Photos(1)

Key Documents

34850-M

Sigma-Aldrich

Acetone

Acetone

suitable for HPLC, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12190000
eCl@ss:
39021201
PubChem Substance ID:
NACRES:
NA.02
grade:
HPLC grade
Assay:
≥99.8%
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

HPLC grade

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

465 °C (869 °F)

expl. lim.

13.2 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.1% water (Karl Fischer)

transmittance

330 nm, ≥15%
335 nm, ≥50%
340 nm, ≥80%
350 nm, ≥98%

refractive index

n20/D 1.359 (lit.)

pH

(5 - 6 at 395 g/l at 20 °C (68 °F))

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

solubility

water: miscible

density

0.791 g/mL at 25 °C (lit.)

application(s)

general analytical

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700°C or 650°C efficiently causes oxidation of acetone.[1] Its Aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions.[2] It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide.[3] Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.[4]

Application

Acetone has been used in the preparation of 13wt% amorphous poly (lactic acid) (PLA) solution, which was required for the preparation of electrospun PLA membranes.[5]
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Other Notes

Important notice
  • The article number 34850-4X2.5L-M will be discontinued. Please order the single bottle 34850-2.5L-M which is physically identical with the same exact specifications.
  • The article number 34850-6X1L-M will be discontinued. Please order the single bottle 34850-1L-M which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gianluigi Luppi et al.
The Journal of organic chemistry, 70(18), 7418-7421 (2005-08-27)
[reaction: see text] The aldol condensation of acetone with several isatins is described. The desired compound was obtained in quantitative yield and with good enantioselectivities up to 77%. The best results were obtained with 10 mol % H-D-Pro-L-beta3-hPhg-OBn as a

Articles

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    High quality solvent

    High quality product which performs well in my system

    Helpful?

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service