Skip to Content
Merck
All Photos(1)

Key Documents

34613

Millipore

Acetonitrile solution

suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10.0

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
technique(s):
HPLC: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg

Product Name

Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg

expl. lim.

8 % (lit.)

technique(s)

HPLC: suitable

bp

81-82 °C (lit.)

mp

-48 °C (lit.)

density

0.782 g/mL at 25 °C

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (CH3CN) is one of the simplest forms of organic nitrile, at room-temperature; it exists as a colorless liquid and has ether like odor. Its properties include melting point, -45.7 °C; boiling point, 82 °C; and density, 0.786 g cm-3. It is produced by dehydration of acetamide and can also be obtained by hydrogenation of a mixture of carbon monoxide and ammonia. Upon hydrolysis, it produces acetic acid and ammonia, whereas upon reduction it gives ethylamine. It is very slightly toxic, though upon metabolism in body gives highly toxic hydrogen cyanide. It is completely miscible with water. Generally it finds application, as an eluant in high-performance liquid chromatography (HPLC).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Yvon et al.
Journal of dairy science, 102(9), 7697-7706 (2019-07-22)
Nutrition plays a crucial role in human gut health through the improvement of gut barrier functionality. Donkey milk represents an interesting source of natural antimicrobial factors such as lysozyme. Recently, anti-inflammatory properties of donkey milk lysozyme activity were described in
Ranen Aviner et al.
PLoS genetics, 11(10), e1005554-e1005554 (2015-10-07)
Studying the complex relationship between transcription, translation and protein degradation is essential to our understanding of biological processes in health and disease. The limited correlations observed between mRNA and protein abundance suggest pervasive regulation of post-transcriptional steps and support the
Seung-Ho Shin et al.
PloS one, 10(9), e0137011-e0137011 (2015-09-04)
Congenital cholesteatomas are epithelial lesions that present as an epithelial pearl behind an intact eardrum. Congenital and acquired cholesteatomas progress quite differently from each other and progress patterns can provide clues about the unique origin and pathogenesis of the abnormality.
Fang Xu et al.
The Plant cell, 27(5), 1547-1562 (2015-05-15)
Nod-like receptors (NLRs) serve as immune receptors in plants and animals. The stability of NLRs is tightly regulated, though its mechanism is not well understood. Here, we show the crucial impact of N-terminal acetylation on the turnover of one plant
Dwight R Stoll et al.
Analytical and bioanalytical chemistry, 407(1), 265-277 (2014-07-30)
In this paper, we describe the findings of a study aimed at assessing the detection sensitivity of comprehensive two-dimensional high-performance liquid chromatography (LCxLC) separation of a degraded active pharmaceutical ingredient (API) with UV absorption as the detection technique. Specifically, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service