Skip to Content
Merck
All Photos(1)

Key Documents

34546

Supelco

2-(Diethylamino)-6-methyl-4-pyrimidinol

PESTANAL®, analytical standard

Synonym(s):

DEAMPY, 2-(Diethylamino)-4-hydroxy-6-methylpyrimidine, 2-(Diethylamino)-6-methyl-1H-pyrimidin-4-one, 2-(Diethylamino)-6-methyl-4(3H)-pyrimidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3O
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

OC1=NC(N(CC)CC)=NC(C)=C1

InChI

1S/C9H15N3O/c1-4-12(5-2)9-10-7(3)6-8(13)11-9/h6H,4-5H2,1-3H3,(H,10,11,13)

InChI key

NQCPECCCWDWTJJ-UHFFFAOYSA-N

General description

2-(Diethylamino)-6-methyl-4-pyrimidinol is a metabolite of pirimiphos-methyl, which is a broad-spectrum organophosphate insecticide used for the protection of food from pests.[1][2][3]

Application

2-(Diethylamino)-6-methyl-4-pyrimidinol may be used as an analytical standard for the determination of the analyte in human biological samples[4][5][1] and fish tissues[6] by liquid chromatography (LC) based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical methods for biological monitoring of exposure to pesticides: a review
Barr DB and Needham LL
Journal of Chromatography. B, Biomedical Applications, 778(1-2), 5-29 (2002)
LC-MS/MS method for the determination of organophosphorus pesticides and their metabolites in salmon and zebrafish fed with plant-based feed ingredients
Garlito B, et al.
Analytical and Bioanalytical Chemistry, 411(27), 7281-7291 (2019)
Distributions and determinants of urinary biomarkers of organophosphate pesticide exposure in a prospective Spanish birth cohort study
Llop S, et al.
Environmental Health : A Global Access Science Source, 16(1), 46-46 (2017)
Comprehensive analytical strategy for biomonitoring of pesticides in urine by liquid chromatography-orbitrap high resolution mass spectrometry
Roca M, et al.
Journal of Chromatography A, 1374(1), 66-76 (2014)
Retrospective analysis of pesticide metabolites in urine using liquid chromatography coupled to high-resolution mass spectrometry
Lopez A, et al.
Talanta, 160, 547-555 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service