Skip to Content
Merck
All Photos(1)

Key Documents

34523

Supelco

Linuron-(methyl-d3, methoxy-d3)

PESTANAL®, analytical standard

Synonym(s):

3-(3,4-Dichlorophenyl)-1-methoxy-d3-1-methyl-d3-urea, Linuron-d6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$723.00

$723.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$723.00

About This Item

Empirical Formula (Hill Notation):
C9D6H4Cl2N2O2
CAS Number:
Molecular Weight:
255.13
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$723.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(NC1=CC(Cl)=C(Cl)C=C1)N(OC([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C9H10Cl2N2O2/c1-13(15-2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)/i1D3,2D3

InChI key

XKJMBINCVNINCA-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service