Skip to Content
Merck
All Photos(1)

Key Documents

34080

Supelco

2,6-Dibromoquinone-4-chloroimide

purum p.a., for the determination of CN-, phenols, for the detection of phosphatase, ≥98.0% (AT)

Synonym(s):

2,6-Dibromo-N-chloro-p-benzoquinoneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Br2ClNO
CAS Number:
Molecular Weight:
299.35
Beilstein:
2094249
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

purum p.a.

Assay

≥98.0% (AT)

quality

for the detection of phosphatase
for the determination of CN-, phenols

technique(s)

UV/Vis spectroscopy: suitable

mp

75-84 °C

storage temp.

2-8°C

SMILES string

Cl\N=C1\C=C(Br)C(=O)C(Br)=C1

InChI

1S/C6H2Br2ClNO/c7-4-1-3(10-9)2-5(8)6(4)11/h1-2H

InChI key

JYWKEVKEKOTYEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to orange-brown on storage

Other Notes

Reagent for the determination of phenols after TLC; Determination of organophosphorus comp.; Determination of cyanide

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Self-react. A - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Funk et al.
J. Planar Chromatogr., 2, 285-285 (1989)
G.G. Guilbault et al
Analytical Chemistry, 37, 1395-1395 (1965)
D. Svobodova et al.
Mikrochimica Acta, 251-251 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service