Skip to Content
Merck
All Photos(1)

Key Documents

33897

Supelco

Thiacloprid-amide

PESTANAL®, analytical standard

Synonym(s):

[3-(6-Chloro-3-pyridylmethyl)thiazolidin-2-ylidene]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClN4OS
CAS Number:
Molecular Weight:
270.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=O)\N=C1/SCCN1Cc2ccc(Cl)nc2

InChI

1S/C10H11ClN4OS/c11-8-2-1-7(5-13-8)6-15-3-4-17-10(15)14-9(12)16/h1-2,5H,3-4,6H2,(H2,12,16)/b14-10-

InChI key

LEZHOZPJYAQQNU-UVTDQMKNSA-N

General description

Thiacloprid-amide is a metabolite of thiacloprid, an insecticide belonging to the class of neonicotinoids.[1] It is primarily used for foliar applications for pest control in arable crops, orchards, vegetables, and other crops.[1]

Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiacloprid-amide may be used as an analytical reference standard for the determination of the analyte in:
  • Green pepper/tomato samples using water based extraction and solid-phase extraction followed by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[2]
  • Honey bees and honey samples using solid-phase extraction followed by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of neonicotinoid insecticides and their metabolites in honey bee and honey by liquid chromatography tandem mass spectrometry.
Gbylik-Sikorska M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 990(7), 132-140 (2015)
Water-based extraction and liquid chromatography-tandem mass spectrometry analysis of neonicotinoid insecticides and their metabolites in green pepper/tomato samples.
Iwafune T, et al.
Journal of Agricultural and Food Chemistry, 62(13), 2790-2796 (2014)
Performance of honey bee colonies under a long-lasting dietary exposure to sublethal concentrations of the neonicotinoid insecticide thiacloprid.
Siede R, et al.
Pest Management Science, 73(7), 1334-1344 (2017)
Kouji H Harada et al.
PloS one, 11(1), e0146335-e0146335 (2016-01-06)
Neonicotinoids, which are novel pesticides, have entered into usage around the world because they are selectively toxic to arthropods and relatively non-toxic to vertebrates. It has been suggested that several neonicotinoids cause neurodevelopmental toxicity in mammals. The aim was to
Marc Le Vée et al.
Journal of biochemical and molecular toxicology, 33(10), e22379-e22379 (2019-08-01)
The interactions of six neonicotinoid pesticides and one neonicotinoid metabolite with drug transporters have been characterized in vitro. Acetamiprid, clothianidin, imidacloprid, nitenpyram, thiacloprid and its metabolite thiacloprid amide, and thiamethoxam, each used at 100 µM, did not impair activity of the

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service