Skip to Content
Merck
All Photos(1)

Documents

33684

Supelco

BDE No 154 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,4′,5,6′-Hexabromodiphenyl ether solution, PBDE 154

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Br6O
CAS Number:
Molecular Weight:
643.58
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1cc(Br)c(Oc2cc(Br)c(Br)cc2Br)c(Br)c1

InChI

1S/C12H4Br6O/c13-5-1-9(17)12(10(18)2-5)19-11-4-7(15)6(14)3-8(11)16/h1-4H

InChI key

VHNPZYZQKWIWOD-UHFFFAOYSA-N

General description

BDE No 154 is a pollutant with low acute toxicity, and belongs to the class of polybrominated diphenyl ethers. It can be commercially used as a flame retardant additive in building materials, textiles, plastics and electronic equipment.

Application

BDE No 154 may be used as an analytical reference standard for the determination of the analyte in:
  • House dust samples using isotope dilution method combined with liquid chromatography coupled to negative ionization atmospheric pressure photoionization tandem mass spectrometry (LC-NI-APPI-MS/MS).
  • Adipose tissue samples using gas chromatography coupled to ion-trap mass spectrometry (GC-ITMS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotope dilution method for determination of polybrominated diphenyl ethers using liquid chromatography coupled to negative ionization atmospheric pressure photoionization tandem mass spectrometry: validation and application to house dust.
Abdallah E-A M, et al.
Analytical Chemistry, 81(17), 7460-7469 (2009)
Development of a gas chromatography/ion trap mass spectrometry based method for the quantification of polybrominated diphenyl ethers and polychlorinated biphenyls in adipose tissue.
Naert C, et al.
Rapid Communications in Mass Spectrometry, 18(19), 2317-2332 (2004)
Luca Maria Chiesa et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(7), 1340-1355 (2018-03-14)
Demand for honey is increasing, especially if it is organic and if its nutritional properties are linked to untreated environments in order to guarantee quality for health. Sources of contamination of honey can be divided into environmental and apicultural. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service