Skip to Content
Merck
All Photos(1)

Key Documents

33588

Supelco

Amidosulfuron

PESTANAL®, analytical standard

Synonym(s):

1-(4,6-Dimethoxypyrimidin-2-yl)-3-(N-mesyl-N-methylsulfamoyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$358.00

$358.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$358.00

About This Item

Empirical Formula (Hill Notation):
C9H15N5O7S2
CAS Number:
Molecular Weight:
369.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$358.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(NC(=O)NS(=O)(=O)N(C)S(C)(=O)=O)n1

InChI

1S/C9H15N5O7S2/c1-14(22(4,16)17)23(18,19)13-9(15)12-8-10-6(20-2)5-7(11-8)21-3/h5H,1-4H3,(H2,10,11,12,13,15)

InChI key

CTTHWASMBLQOFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amidosulfuron is a selective, sulfonylurea herbicide employed for the control of many grasses and broadleaf weeds in crop protection of vines, rice, citrus, corn, potatoes and tomatoes [1]

Application

Amidosulfuron may be used as a reference standard for the determination of the analyte in surface waters using high-performance liquid chromatography with ultraviolet detection (HPLC-UV) and mass spectrometry (MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura R Davies et al.
Pest management science, 76(7), 2473-2482 (2020-02-16)
Anisantha and Bromus spp. are widespread and difficult to control, potentially due to the evolution of herbicide resistance. In this study, UK populations of four brome species have been tested for the early development of resistance to acetolactate synthase (ALS)-inhibiting
Ana Zabalza et al.
Plants (Basel, Switzerland), 9(9) (2020-09-20)
Quinate (1,3,4,5-tetrahydroxycyclohexanecarboxylate) is a compound synthesized in plants through a side-branch of the shikimate biosynthesis pathway, which is accumulated after glyphosate and acetolactate synthase inhibiting herbicides (ALS-inhibitors) and has phytotoxic potential. The objective of this study was to evaluate the
Petr Vítek et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 170, 234-241 (2016-07-28)
The effects of herbicides from three mode-of-action groups - inhibitors of protoporphyrinogen oxidase (carfentrazone-ethyl), inhibitors of carotenoid biosynthesis (mesotrione, clomazone, and diflufenican), and inhibitors of acetolactate synthase (amidosulfuron) - were studied in sunflower plants (Helianthus annuus). Raman spectroscopy, chlorophyll fluorescence
A Gimeno et al.
Journal of applied microbiology, 129(3), 680-694 (2020-03-17)
To evaluate biological control agents (BCAs) against Fusarium graminearum on infected maize stalks as a means to reduce Fusarium head blight (FHB) in subsequently grown wheat. In the laboratory, BCAs were applied against F. graminearum on maize stalk pieces. Clonostachys
HPLC-UV and HPLC-MSn multiresidue determination of amidosulfuron, azimsulfuron, nicosulfuron, rimsulfuron, thifensulfuron methyl, tribenuron methyl and azoxystrobin in surface waters.
Polati S, et al.
Analytica Chimica Acta, 579(2), 146-151 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service