Skip to Content
Merck
All Photos(1)

Key Documents

33495

Supelco

Famoxadone solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

3-Anilino-5-methyl-5-(4-phenoxyphenyl)oxazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$122.00

$122.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$122.00

About This Item

Empirical Formula (Hill Notation):
C22H18N2O4
CAS Number:
Molecular Weight:
374.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$122.00


Estimated to ship on19 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

O=C1OC(C(C=C2)=CC=C2OC3=CC=CC=C3)(C)C(N1NC4=CC=CC=C4)=O

InChI

1S/C22H18N2O4/c1-22(16-12-14-19(15-13-16)27-18-10-6-3-7-11-18)20(25)24(21(26)28-22)23-17-8-4-2-5-9-17/h2-15,23H,1H3

InChI key

PCCSBWNGDMYFCW-UHFFFAOYSA-N

General description

Famoxadone belongs to the oxazolidinedione class of fungicides used to control broad spectrum of fungal diseases in agricultural crops.[1]

Application

Famoxadone may be used as a reference standard for the determination of the analyte in wine and grape samples using gas chromatography (GC) coupled to electron capture (EC) and mass spectrometric (MS) methods of detection respectively.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guofeng Xu et al.
Journal of separation science, 41(20), 3871-3880 (2018-08-24)
Famoxadone is a widely used chiral fungicide on tomato, apple, and grape. But it is still being employed as a racemic mixture without distinguishing the difference between enantiomers, which often leads to its inaccurate risk assessment. In this study, a
A comparison of a gas chromatographic with electron-capture detection and a gas chromatographic with mass spectrometric detection screening methods for the analysis of famoxadone in grapes and wines.
Abreu MDS, et al.
Journal of Chromatography A, 1103(2), 362-367 (2006)
Yongbo Cui et al.
PloS one, 12(7), e0181075-e0181075 (2017-07-13)
The aim of this study was to elucidate the molecular mechanisms by which food-derived casein glycomacropeptide (CGMP) maintains internal homeostasis in the intestinal mucosa and to investigate the effects of CGMP on the intestinal mucosal immunological barrier and related signal
Mohd Aamir et al.
Frontiers in pharmacology, 9, 1038-1038 (2018-11-09)
Vascular wilt of tomato caused by Fusarium oxysporum f.sp. lycopersici (FOL) is one of the most devastating diseases, that delimits the tomato production worldwide. Fungal short-chain dehydrogenases/reductases (SDRs) are NADP(H) dependent oxidoreductases, having shared motifs and common functional mechanism, have
Marcin Sarewicz et al.
Biochemistry, 52(37), 6388-6395 (2013-08-15)
Efficient energy conversion often requires stabilization of one-electron intermediates within catalytic sites of redox enzymes. While quinol oxidoreductases are known to stabilize semiquinones, one of the famous exceptions includes the quinol oxidation site of cytochrome bc1 (Qo), for which detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service