Skip to Content
Merck
All Photos(1)

Key Documents

33028

Supelco

Cyanofenphos solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14NO2PS
CAS Number:
Molecular Weight:
303.32
Beilstein:
2945855
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=S)(Oc1ccc(cc1)C#N)c2ccccc2

InChI

1S/C15H14NO2PS/c1-2-17-19(20,15-6-4-3-5-7-15)18-14-10-8-13(12-16)9-11-14/h3-11H,2H2,1H3

InChI key

LRNJHZNPJSPMGK-UHFFFAOYSA-N

General description

Cyanofenphos belongs to the class of phosphonothioate insecticides. It is used to control various insect pests in agriculture.

Application

Cyanofenphos may be used as an analytical reference standard for the determination of the analyte in:
  • Nutraceutical products by liquid chromatography Orbitrap-mass spectrometry (LC-Orbitrap-MS).
  • Animal samples by gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Agricultural products by GC-MS.
  • Fruits and vegetables by GC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-pressure liquid chromatography of neurotoxic phenylphosphonothioate esters and related compounds.
J M Lasker et al.
Analytical biochemistry, 109(2), 369-375 (1980-12-01)
Determination of toxic substances, pesticides and mycotoxins, in ginkgo biloba nutraceutical products by liquid chromatography Orbitrap-mass spectrometry.
Martinez-Dominguez G, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 118(2), 124-130 (2015)
N Konno et al.
The Tohoku journal of experimental medicine, 142(2), 155-163 (1984-02-01)
Cyanofenphos (Surecide, O-ethyl O-4-cyanophenyl phenylphosphonothioate) is an organophosphorus pesticide. The protective effect against the near lethal dose, 140% the LD50, of formulated cyanofenphos developed in hens pretreated daily with a small dosage of 1.4% the LD50. The oral LD50 for
Delayed neuropathy in sheep by the phosphonothioate insecticide cyanofenphos.
El-Sebae AH, et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 14(3), 247-263 (1979)
E Enan et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 22(2), 149-170 (1987-04-01)
Male Baladi rabbits were acutely and sub-chronically intoxicated with cyanofenphos and profenophos. The levels of cholesterol, triglycerides, B-lipoproteins and total proteins were determined in the serum, brain, spinal cord and sciatic nerve of rabbits. Moreover, the activities of alkaline phosphatase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service