Skip to Content
Merck
All Photos(1)

Key Documents

32702

Supelco

4-(1-Ethyl-1-methylhexyl)phenol

analytical standard

Synonym(s):

3M3-Nonylphenol isomer, 4-(3-Methyl-3-octyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein:
10343926
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

−20°C

SMILES string

CCCCCC(C)(CC)c1ccc(O)cc1

InChI

1S/C15H24O/c1-4-6-7-12-15(3,5-2)13-8-10-14(16)11-9-13/h8-11,16H,4-7,12H2,1-3H3

InChI key

QQKQQJZGUCFUPP-UHFFFAOYSA-N

General description

4-(1-Ethyl-1-methylhexyl)phenol is one of the isomers of 4-nonylphenol which is regarded as an environmental endocrine disruptor.

Application

4-(1-Ethyl-1-methylhexyl)phenol has been used as an analytical reference standard for the determination of the analyte in wastewater and surface water samples by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
It may be used as an analytical standard for the determination of the analyte in river sediment samples by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isomer-specific biodegradation of nonylphenol in river sediments and structure-biodegradability relationship.
Lu L and Gan J
Environmental Science & Technology, 48(2), 1008-1014 (2014)
Syntheses and estrogenic activity of 4-nonylphenol isomers.
Uchiyama T, et al.
Chemosphere, 73(1), S60-S65 (2008)
Liquid chromatographic-tandem mass spectrometric method for the simultaneous determination of alkylphenols polyethoxylates, alkylphenoxy carboxylates and alkylphenols in wastewater and surface-water.
Ciofi L, et al.
Journal of Chromatography A, 1362(1), 75-88 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service