Skip to Content
Merck
All Photos(2)

Key Documents

32434

Supelco

TEPP

PESTANAL®, analytical standard

Synonym(s):

Tetraethyl pyrophosphate, Tetraethyl diphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$167.00

$167.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$167.00

About This Item

Empirical Formula (Hill Notation):
C8H20O7P2
CAS Number:
Molecular Weight:
290.19
Beilstein:
1714017
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$167.00


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)OP(=O)(OCC)OCC

InChI

1S/C8H20O7P2/c1-5-11-16(9,12-6-2)15-17(10,13-7-3)14-8-4/h5-8H2,1-4H3

InChI key

IDCBOTIENDVCBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TEPP (Tetraethyl pyrophosphate, Pestanal) is highly toxic in nature both by skin absorption and inhalation. It is an organic phosphate pesticide, acting acts as an inhibitor of cholinesterase. It is soluble in water and is slowly decomposed by water. It may be found as a dry mixture or in the liquid state.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversal of the actions of tetraethyl pyrophosphate in surviving mammalian tissue.
B B ROY et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 89(2), 255-258 (1955-06-01)
Studies on the mechanism of action of DFP and TEPP.
B P McNAMARA et al.
The Journal of pharmacology and experimental therapeutics, 110(2), 232-240 (1954-02-01)
Tetraethyl pyrophosphate in the myasthenia gravis.
O J NOVOTA
Missouri medicine, 49(2), 133-134 (1952-02-01)
J U Patel et al.
Journal of pharmaceutical sciences, 83(10), 1477-1481 (1994-10-01)
An O-(saccharinylmethyl) prodrug was synthesized to improve the poor oral potency of the phenolic drug 17 beta-estradiol. This O-(imidomethyl) type of prodrug was designed to undergo chemical hydrolysis and to be a poor substrate for enzymatic hydrolysis. At 37 degrees
N S L Perry et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 9(1), 48-51 (2002-04-03)
The essential oil of Salvia lavandulaefolia at two dosage levels was administered orally to rats for five days. Choline esterase activity was measured post mortem in three areas of the brain, both in the absence and presence of TEPP, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service