Skip to Content
Merck
All Photos(1)

Key Documents

32391

Supelco

Flucarbazone sodium

PESTANAL®, analytical standard

Synonym(s):

Flucarbazone sodium salt, 4,5-Dihydro-3-methoxy-4-methyl-5-oxo-N-(2-trifluoromethoxyphenylsulfonyl)-1H-1,2,4-triazole-1-carboxamide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F3N4NaO6S
CAS Number:
Molecular Weight:
418.28
UNSPSC Code:
12352202

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

InChI

1S/C12H11F3N4O6S.Na/c1-18-10(24-2)16-19(11(18)21)9(20)17-26(22,23)8-6-4-3-5-7(8)25-12(13,14)15;/h3-6H,1-2H3,(H,17,20);/q;+1/p-1

InChI key

UOUXAYAIONPXDH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service