Skip to Content
Merck
All Photos(1)

Key Documents

32378

Supelco

Methapyrilene-(dimethyl-d6) fumarate salt

VETRANAL®, analytical standard

Synonym(s):

N,N-Dimethyl-d6-N′-(2-pyridyl)- N′-(2-thienylmethyl)-1,2-ethandiamine fumarate salt (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D6H13N3S · C4H4O4
CAS Number:
Molecular Weight:
383.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

O=C(O)/C=C/C(O)=O.[2H]C([2H])([2H])N(C([2H])([2H])[2H])CCN(CC1=CC=CS1)C2=NC=CC=C2

InChI

1S/C14H19N3S.C4H4O4/c1-16(2)9-10-17(12-13-6-5-11-18-13)14-7-3-4-8-15-14;5-3(6)1-2-4(7)8/h3-8,11H,9-10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/i1D3,2D3;

InChI key

GYIUBPDECOMZDC-FEYLKBJGSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service